Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-tert-butyl-1,4-benzoquinone | 3602-55-9 | MFCD00666928 | 99.0% | 164.20 g/mol | C10H12O2 | 10 G
SDP

Catalog No. 5000300150
Encompass_Preferred

2-tert-butyl-1,4-benzoquinone is a tert-butyl substituted 1,4-benzoquinone supplied for laboratory research. It is reported as an electrophilic metabolite of butylated hydroxyanisole and as an oxidation product of 2-tert-butylhydroquinone. The material is provided as a light yellow solid and is characterized by a molecular formula of C10H12O2, a molecular weight of 164.20 g/mol, and manufacturer-reported GC purity of 99%.

  • Light yellow solid appearance.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • High purity: 99.0% (GC) per certificate of analysis.
  • Analytical data includes GC purity, water (KF), residue on ignition, and 1H NMR confirmation.
  • Storage recommendations provided for powder and for solutions to maintain stability.
  • Available in multiple laboratory-scale package sizes.

Catalog No. 50-003-00150 Supplier Medchemexpress LLC Supplier No. HYW01718710G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-tert-butyl-1,4-benzoquinone is a tert-butyl substituted 1,4-benzoquinone supplied for laboratory research. It is reported as an electrophilic metabolite of butylated hydroxyanisole and as an oxidation product of 2-tert-butylhydroquinone. The material is provided as a light yellow solid and is characterized by a molecular formula of C10H12O2, a molecular weight of 164.20 g/mol, and manufacturer-reported GC purity of 99%.

  • Light yellow solid appearance.
  • Molecular formula C10H12O2 and molecular weight 164.20 g/mol.
  • High purity: 99.0% (GC) per certificate of analysis.
  • Analytical data includes GC purity, water (KF), residue on ignition, and 1H NMR confirmation.
  • Storage recommendations provided for powder and for solutions to maintain stability.
  • Available in multiple laboratory-scale package sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.