Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-tert-butyl-1,4-benzoquinone | 3602-55-9 | MFCD00666928 | 99.0% | 164.20 g/mol | C10H12O2 | 250 G
SDP

Catalog No. 5000302701
Encompass_Preferred

2-tert-butyl-1,4-benzoquinone (CAS 3602-55-9) is an electrophilic benzoquinone and an oxidation/metabolite product of 2-tert-butylhydroquinone and butylated hydroxyanisole. It is supplied as a research-grade solid reagent for studies of oxidative metabolism and electrophilic reactivity.

  • High purity: 99.0% as supplied.
  • Physical form: solid, light yellow to yellow.
  • Chemical data: formula C10H12O2; molecular weight 164.20 g/mol.
  • Typical uses: studies of metabolism, electrophilic reactivity, and chemical synthesis.
  • Packaging: available in multiple sizes, including 250 g.

Catalog No. 50-003-02701 Supplier Medchemexpress LLC Supplier No. 250GHYW017187
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-tert-butyl-1,4-benzoquinone (CAS 3602-55-9) is an electrophilic benzoquinone and an oxidation/metabolite product of 2-tert-butylhydroquinone and butylated hydroxyanisole. It is supplied as a research-grade solid reagent for studies of oxidative metabolism and electrophilic reactivity.

  • High purity: 99.0% as supplied.
  • Physical form: solid, light yellow to yellow.
  • Chemical data: formula C10H12O2; molecular weight 164.20 g/mol.
  • Typical uses: studies of metabolism, electrophilic reactivity, and chemical synthesis.
  • Packaging: available in multiple sizes, including 250 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.