Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(tert-butylamino)acetic acid hydrochloride | 6939-23-7 | MFCD03788441 | ≥98.0% | 167.63 | C6H14ClNO2 | 500 G
SDP

Catalog No. 5000208732
Encompass_Preferred

2-(tert-butylamino)acetic acid hydrochloride is the hydrochloride salt of 2-(tert-butylamino)acetic acid, a glycine derivative used in biochemical and pharmacological research. It is supplied for research use only and is commonly used as an amino acid derivative in laboratory studies.

  • Glycine derivative used in biochemical research.
  • Cas number 6939-23-7.
  • Molecular formula C6H14ClNO2.
  • Molecular weight 167.63 g/mol.
  • Purity ≥98.0%.
  • Available in 25 g, 100 g, and 500 g packages.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-08732 Supplier Medchemexpress LLC Supplier No. HYW040723500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(tert-butylamino)acetic acid hydrochloride is the hydrochloride salt of 2-(tert-butylamino)acetic acid, a glycine derivative used in biochemical and pharmacological research. It is supplied for research use only and is commonly used as an amino acid derivative in laboratory studies.

  • Glycine derivative used in biochemical research.
  • Cas number 6939-23-7.
  • Molecular formula C6H14ClNO2.
  • Molecular weight 167.63 g/mol.
  • Purity ≥98.0%.
  • Available in 25 g, 100 g, and 500 g packages.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.