Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(tert-butylamino)acetic acid hydrochloride | 6939-23-7 | MFCD03788441 | >98.0% | 167.63 g/mol | C6H14ClNO2 | 100 G
SDP

Catalog No. 5000209380
Encompass_Preferred

2-(tert-butylamino)acetic acid hydrochloride is the hydrochloride salt of a tert-butylamino-substituted glycine derivative supplied as a white to off-white solid for research use. It is commonly used as an amino acid derivative and reagent in peptide synthesis and related biochemical applications.

  • White to off-white solid with >98.0% purity.
  • Molecular formula C6H14ClNO2 and molecular weight 167.63 g/mol.
  • CAS number 6939-23-7 for unambiguous identification.
  • Typically available in laboratory pack sizes such as 25 g, 100 g, and 500 g.
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years); follow recommended storage for solutions.
  • Intended for research use in peptide synthesis and biochemical studies.

Catalog No. 50-002-09380 Supplier Medchemexpress LLC Supplier No. HYW040723100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(tert-butylamino)acetic acid hydrochloride is the hydrochloride salt of a tert-butylamino-substituted glycine derivative supplied as a white to off-white solid for research use. It is commonly used as an amino acid derivative and reagent in peptide synthesis and related biochemical applications.

  • White to off-white solid with >98.0% purity.
  • Molecular formula C6H14ClNO2 and molecular weight 167.63 g/mol.
  • CAS number 6939-23-7 for unambiguous identification.
  • Typically available in laboratory pack sizes such as 25 g, 100 g, and 500 g.
  • Store powder at -20°C (up to 3 years) or 4°C (up to 2 years); follow recommended storage for solutions.
  • Intended for research use in peptide synthesis and biochemical studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.