Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(tert-Butylamino)ethanol, TCI America™
SDP

Catalog No. B1101500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1101500G 500 g
B110125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1101500G Supplier TCI America Supplier No. B1101500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4620-70-6
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00002837
InChI Key IUXYVKZUDNLISR-UHFFFAOYSA-N
Synonym 2-tert-butylamino ethanol, n-tert-butylethanolamine, tert-butylaminoethanol, t-butylethanolamine, ethanol, 2-tert-butylamino, ethanol, 2-1,1-dimethylethyl amino, n-t-butylethanolamine, 2-t-butylamino ethanol, 2-1,1-dimethylethyl amino ethanol, .beta.-tert-butylaminoethanol
PubChem CID 78361
IUPAC Name 2-(tert-butylamino)ethan-1-ol
SMILES CC(C)(C)NCCO

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 177°C
UN Number 2923
Quantity 500 g
Formula Weight 117.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-(tert-Butylamino)ethanol
EINECSNumber (2)-0295
RTECSNumber KK7177500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.