Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylcyclohexanone 97%, Thermo Scientific™

Catalog No. AC217761000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC217761000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC217761000 Supplier Thermo Scientific Chemicals Supplier No. 217761000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1728-46-7
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
InChI Key ZRYDPLOWJSFQAE-UHFFFAOYSA-N
Synonym 2-tert-butyl cyclohexanone, 2-tert-butylcyclohexanone, 2-t-butylcyclohexanone, cyclohexanone, 2-1,1-dimethylethyl, ortho-tert-butylcyclohexanone, acmc-20ag5y, 2-tert-butyl-cyclohexanone, 2-tert. butyl-cyclohexanone, dsstox_cid_27081, dsstox_rid_82093
PubChem CID 102681
IUPAC Name 2-tert-butylcyclohexan-1-one
SMILES CC(C)(C)C1CCCCC1=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Green-Yellow
Infrared Spectrum Authentic
Linear Formula (CH3)3CC6H9(=O)
Refractive Index 1.4560 to 1.4580 (20°C, 589nm)
Quantity 100 g
Beilstein 07, III, 143
Formula Weight 154.25
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-tert-Butylcyclohexanone, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.