Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Tert-Butylimino-2-Diethylamino-1,3-Dimethylperhydro-1,3,2-Diazaphosphorine

Catalog No. 200255ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-025-5ML 5 mL
20-025-25ML 25 mL
20-025-1ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 20-025-5ML Supplier Sigma Aldrich Supplier No. 200255ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine (BEMP) is a phosphazene base. BEMP participates in the mild base catalyzed nucleophilic ring opening of N-sulfonyl aziridines. BEMP supported on polystyrene (PS-BEMP) has been reported as an efficient catalyst for the ring-opening of epoxides with phenols. BEMP is about 2000 times more basic and also much more sterically hindered than DBU (l,8-diazabicyclo[5.4.0]undecene).Phosphazene Bases: a Family of Extremely Strong, Non-Ionic, Non-Charged Nitrogen-Bases

Specifications

CAS 98015-45-3
Density 0.948 g/mL at 25°C
Boiling Point 74°C (0.03 mmHg, lit.)
Molecular Formula C13H31N4P
Linear Formula C13H31N4P
Refractive Index n20/D 1.477; n20/D 1.477
Quantity 5 mL
MDL Number MFCD00043143
Synonym BEMP
Molecular Weight (g/mol) 274.39 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.