Learn More
Sigma Aldrich 2-Tert-Butylimino-2-Diethylamino-1,3-Dimethylperhydro-1,3,2-Diazaphosphorine, Polymer-Bound
Catalog No. 5364905G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 g
Catalog No. Quantity
53-649-05G 5 g
1 options
Danger
Catalog No. 53-649-05G Supplier Sigma Aldrich Supplier No. 5364905G
Add to Cart
Add to Cart
Danger
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Polymer-supported base used for deprotonation and N-alkylation of weakly acidic heterocycles.
Specifications
CAS 1173023-01-2
Quantity 5 g
Synonym Benzyloxybenzyl chloride, polymer-bound; Wang chlorinated resin; [4-(Chloromethyl)phenoxymethyl]polystyrene cross-linked with divinylbenzene
Biological Activity Respiratory System