Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Tert-Butylimino-2-Diethylamino-1,3-Dimethylperhydro-1,3,2-Diazaphosphorine Solution

Catalog No. 72841125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-841-125ML 25 mL
72-841-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-841-125ML Supplier Sigma Aldrich Supplier No. 72841125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Tert-Butylimino-2-Diethylamino-1,3-Dimethylperhydro-1,3,2-Diazaphosphorine Solution

Specifications

CAS 98015-45-3
Molecular Formula C13H31N4P
Linear Formula C13H31N4P
MDL Number MFCD00043143
Quantity 25 mL
Synonym BEMP
Molecular Weight (g/mol) 274.39 g/mol
Biological Activity Central Nervous System, Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.