Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylpyrazine 97.0+%, TCI America™
SDP

Catalog No. B33941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B33941G Supplier TCI America Supplier No. B33941G

Chemical Identifiers

CAS 32741-11-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00069193
InChI Key YNQZVOWNJKASKQ-UHFFFAOYSA-N
Synonym t-butylpyrazine, pyrazine, 1,1-dimethylethyl, pyrazine, 2-1,1-dimethylethyl, tert-butylpyrazine, 1,1-dimethylethyl pyrazine, 2-t-butylpyrazine, acmc-209hv7
PubChem CID 122943
IUPAC Name 2-tert-butylpyrazine
SMILES CC(C)(C)C1=CN=CC=N1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 136.20
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-tert-Butylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.