Learn More
Medchemexpress LLC 2-(Tetradecylthio)acetic acid | 2921-20-2 | 99.8% | 288.49 | 25 MG
SDP
Catalog No. 5000352486
Encompass_Preferred
2-(Tetradecylthio)acetic acid is a pan-peroxisome proliferator activated receptor (pan-PPAR) activator that induces hypolipidemia. It effectively reduces plasma lipids and enhances hepatic fatty acid oxidation in rodents. The compound also promotes the increased expression of genes crucial for fatty acid uptake, activation, accumulation, and oxidation.
  • Acts as a pan-peroxisome proliferator activated receptor (pan-PPAR) activator.
  • Induces hypolipidemia.
  • Reduces plasma lipids.
  • Enhances hepatic fatty acid oxidation in rodents.
  • Increases the expression of genes involved in fatty acid uptake, activation, accumulation, and oxidation.
Catalog No. 50-003-52486 Supplier Medchemexpress LLC Supplier No. 25MGHY139040
2-(Tetradecylthio)acetic acid is a pan-peroxisome proliferator activated receptor (pan-PPAR) activator that induces hypolipidemia. It effectively reduces plasma lipids and enhances hepatic fatty acid oxidation in rodents. The compound also promotes the increased expression of genes crucial for fatty acid uptake, activation, accumulation, and oxidation.
  • Acts as a pan-peroxisome proliferator activated receptor (pan-PPAR) activator.
  • Induces hypolipidemia.
  • Reduces plasma lipids.
  • Enhances hepatic fatty acid oxidation in rodents.
  • Increases the expression of genes involved in fatty acid uptake, activation, accumulation, and oxidation.