Learn More
2-Tetrahydropyran-4-ylethanol, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 1 g |
| IUPAC Name | 2-(oxan-4-yl)ethanol |
| Formula Weight | 130.19 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Tetrahydropyran-4-ylethanol |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: Harmful in contact with skin. Causes serious eye irritation. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement: IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.