Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Tetrahydropyran-4-ylethanol, 97%, Thermo Scientific™

Catalog No. CC67409DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC67409DE 5 g
CC67409CB 250 mg
CC67409DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC67409DE Supplier Thermo Scientific Chemicals Supplier No. CC67409DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4677-18-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00129068
InChI Key XZXZZACRGBBWTQ-UHFFFAOYSA-N
Synonym 2-tetrahydro-2h-pyran-4-yl ethanol, 2-oxan-4-yl ethan-1-ol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2-oxan-4-yl ethanol, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran, 2h-pyran-4-ethanol, tetrahydro, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-ylethanol
PubChem CID 17750944
SMILES OCCC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 2-(oxan-4-yl)ethan-1-ol
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 2-Tetrahydropyran-4-ylethanol
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.