Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Tetrahydropyran-4-yloxy)benzoyl chloride, 97%, Thermo Scientific™

Catalog No. CC52202CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC52202CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC52202CB Supplier Thermo Scientific Chemicals Supplier No. CC52202CB

missing translation for 'validMainframeMsg'

Specifications

CAS 906352-68-9
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H13ClO3
Quantity 250mg
MDL Number MFCD09025870
Synonym 2-oxan-4-yloxy benzoyl chloride, 2-tetrahydro-2h-pyran-4-yloxy benzoyl chloride, 2-tetrahydropyran-4-yloxy benzoyl chloride, 2-tetrahydro-2h-pyran-4-yl oxy benzoyl chloride, 2-oxan-4-yl oxy benzoyl chloride, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoyl chloride, benzoyl chloride,2-tetrahydro-2h-pyran-4-yl oxy
InChI Key FYJPOEFQSOXMMO-UHFFFAOYSA-N
SMILES ClC(=O)C1=CC=CC=C1OC1CCOCC1
IUPAC Name 2-(oxan-4-yloxy)benzoyl chloride
Molecular Weight (g/mol) 240.68
PubChem CID 24229514
Formula Weight 240.68
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.