Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 2-Thenoyltrifluoroacetone, 326-91-0, MFCD00005445, 25g

Catalog No. 500119150
Encompass_Preferred

Molecular Formula C8H5F3O2S, Purity 99%, Molecular Weight 222.19, Melting Point 40-44° Boiling Point 96-98/8mm

Catalog No. 50-011-9150 Supplier Matrix Scientific Supplier No. 00637025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C8H5F3O2S, Purity 99%, Molecular Weight 222.19, Melting Point 40-44° Boiling Point 96-98/8mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.