Learn More
Medchemexpress LLC 2-Thiabicyclo[3.1.0]hexane-4,6-dicarboxylic acid, 4-amino-, 2,2-dioxide, (1R,4S,5S,6S)- | 635318-11-5 | ≥98.0% | C7H9NO6S | 100 MG
SDP
Catalog No. 5000358883
Encompass_Preferred
LY404039 is a potent, selective, and orally active mGluR2 and mGluR3 agonist. It exhibits >100-fold selectivity for mGluR2/3 over other receptors/transporters and has antipsychotic and anxiolytic effects.
  • Potent, selective, and orally active mGluR2 and mGluR3 agonist
  • Ki values of 149 nM and 92 nM for mGluR2 and mGluR3
  • Exhibits >100-fold selectivity for mGluR2/3 over other receptors/transporters
  • Has antipsychotic effects
  • Has anxiolytic effects
Catalog No. 50-003-58883 Supplier Medchemexpress LLC Supplier No. HY50906100MG
LY404039 is a potent, selective, and orally active mGluR2 and mGluR3 agonist. It exhibits >100-fold selectivity for mGluR2/3 over other receptors/transporters and has antipsychotic and anxiolytic effects.
  • Potent, selective, and orally active mGluR2 and mGluR3 agonist
  • Ki values of 149 nM and 92 nM for mGluR2 and mGluR3
  • Exhibits >100-fold selectivity for mGluR2/3 over other receptors/transporters
  • Has antipsychotic effects
  • Has anxiolytic effects