Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thiazolecarboxaldehyde

Catalog No. 4224601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-246-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-246-01G Supplier Sigma Aldrich Supplier No. 4224601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
  • Sigma-aldrich does not collect analytical data for this product
  • Buyer assumes responsibility to confirm product identity and/or purity
  • All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise.product of chembridge corp

Specifications

CAS 10200-59-6
Density 1.288 g/mL (at 25°C (literature))
Boiling Point 61°C to 63°C (15 mmHg)
Molecular Formula C4H3NOS
Refractive Index n20/D 1.574 (literature)
Linear Formula C4H3NOS
MDL Number MFCD00142924
Quantity 1 g
Molecular Weight (g/mol) 113.14
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.