Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Thiazolemethanol, α-[2-[1-(2-amino-5-chloro-4-pyrimidinyl)-2,3-dihydro-1H-indol-6-yl]ethynyl]-α-methyl | 1202764-53-1 | 99.4% | 50 MG
SDP

Catalog No. 5000234346
Encompass_Preferred

B022 is a potent, selective small-molecule inhibitor of NF-κB-inducing kinase (NIK) used for research applications; it contains an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is supplied as a solid with reported high purity.

  • Potent, selective NIK inhibitor (Ki = 4.2 nM; IC50 ≈ 15.1 nM).
  • Contains an alkyne group suitable for click chemistry (CuAAC).
  • High reported purity (99.37%).
  • Molecular weight 397.88; formula C19H16ClN5OS.
  • Soluble in DMSO (250 mg/mL) with recommended formulation options for in vivo use.
  • Stable when stored as powder at -20°C or 4°C per manufacturer guidance.

Catalog No. 50-002-34346 Supplier Medchemexpress LLC Supplier No. HY12050150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

B022 is a potent, selective small-molecule inhibitor of NF-κB-inducing kinase (NIK) used for research applications; it contains an alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (click chemistry) and is supplied as a solid with reported high purity.

  • Potent, selective NIK inhibitor (Ki = 4.2 nM; IC50 ≈ 15.1 nM).
  • Contains an alkyne group suitable for click chemistry (CuAAC).
  • High reported purity (99.37%).
  • Molecular weight 397.88; formula C19H16ClN5OS.
  • Soluble in DMSO (250 mg/mL) with recommended formulation options for in vivo use.
  • Stable when stored as powder at -20°C or 4°C per manufacturer guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.