Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thienylzinc Bromide Solution

Catalog No. 49941250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-941-250ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 49-941-250ML Supplier Sigma Aldrich Supplier No. 49941250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Thienylzinc bromide can be used as:A starting material in the synthesis of heteroaryl sulfonamides by reacting with 2,4,6-trichlorophenyl chlorosulfate.A substrate in the synthesis of naphthacenodithiophene derived polymers, which are used in transistor and solar cell devices.An intermediate in the preparation of isoquinoline derived AICARFT inhibitors.Product of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 79973-42-5
Melting Point 305°C
Molecular Formula C20H30CoF6P
Linear Formula C20H30CoF6P
MDL Number MFCD00269818
Quantity 1 g
Synonym Decamethylcobaltocenium hexafluorophosphate
Molecular Weight (g/mol) 474.35 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.