Learn More
Medchemexpress LLC 2-thio-PAF | 96801-55-7 | 99.0% | 539.75 | C26H54NO6PS | 5 MG
SDP
Catalog No. 5000212725
Encompass_Preferred
2-Thio-PAF is a synthetic analogue of platelet-activating factor (PAF) used as a biochemical reagent. It functions as a substrate in colorimetric assays to measure PAF acetylhydrolase activity and is supplied as a high-purity small-molecule solution for research and assay applications.
  • High purity: 99.0% suitable for biochemical assays.
  • Molecular weight 539.75 g/mol for accurate stoichiometry.
  • Designed as a colorimetric substrate for PAF acetylhydrolase assays.
  • Available in small pack sizes, including 5 mg.
  • Datasheet, certificate of analysis, and safety data sheet available.
Catalog No. 50-002-12725 Supplier Medchemexpress LLC Supplier No. HY1012635MG
2-Thio-PAF is a synthetic analogue of platelet-activating factor (PAF) used as a biochemical reagent. It functions as a substrate in colorimetric assays to measure PAF acetylhydrolase activity and is supplied as a high-purity small-molecule solution for research and assay applications.
  • High purity: 99.0% suitable for biochemical assays.
  • Molecular weight 539.75 g/mol for accurate stoichiometry.
  • Designed as a colorimetric substrate for PAF acetylhydrolase assays.
  • Available in small pack sizes, including 5 mg.
  • Datasheet, certificate of analysis, and safety data sheet available.