Learn More
Medchemexpress LLC 2-Thio-PAF | 96801-55-7 | 99.0% | 539.75 g/mol | C26H54NO6PS | 10 MG
SDP
Catalog No. 5000212861
Encompass_Preferred
2-Thio-PAF is a synthetic analog of platelet-activating factor used as a chromogenic substrate in colorimetric assays for PAF acetylhydrolases. The compound contains a thioester at the sn-2 position, which provides increased stability while allowing enzymatic cleavage for activity measurements in biochemical and enzymology studies.
  • Used as a chromogenic substrate for PAF acetylhydrolase assays.
  • Thioester modification increases stability compared with native PAF.
  • High purity supports reproducible enzymatic measurements.
  • Suitable for biochemical and enzymology research applications.
  • Recommended storage at -20 °C for solution stability.
Catalog No. 50-002-12861 Supplier Medchemexpress LLC Supplier No. HY10126310MG
2-Thio-PAF is a synthetic analog of platelet-activating factor used as a chromogenic substrate in colorimetric assays for PAF acetylhydrolases. The compound contains a thioester at the sn-2 position, which provides increased stability while allowing enzymatic cleavage for activity measurements in biochemical and enzymology studies.
  • Used as a chromogenic substrate for PAF acetylhydrolase assays.
  • Thioester modification increases stability compared with native PAF.
  • High purity supports reproducible enzymatic measurements.
  • Suitable for biochemical and enzymology research applications.
  • Recommended storage at -20 °C for solution stability.