Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneacetic acid, 98%

Catalog No. AC187730250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187730250 25 g Plastic bottle
AC187730050 5 g Glass Bottle
AC187731000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC187730250 Supplier Thermo Scientific Chemicals Supplier No. 187730250
Only null left
Add to Cart

CAS: 1918-77-0 | C6H6O2S | 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-77-0
MDL Number MFCD00005458
InChI Key SMJRBWINMFUUDS-UHFFFAOYSA-N
Synonym 2-thiopheneacetic acid, 2-thienylacetic acid, thiopheneacetic acid, 2-thiophen-2-yl acetic acid, thiophene-2-acetic acid, 2-2-thienyl acetic acid, 2-thiophene acetic acid, thiophen-2-ylacetic acid, thien-2-yl acetic acid, 2-carboxymethyl thiophene
PubChem CID 15970
ChEBI CHEBI:45807
IUPAC Name 2-thiophen-2-ylacetic acid
SMILES C1=CSC(=C1)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 61°C to 65°C
Color Brown
Boiling Point 160°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C6H6O2S
Quantity 25 g
Beilstein 18, 293
Molecular Weight (g/mol) 142.17
Formula Weight 142.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiopheneacetic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-639-8
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.