Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiopheneacetyl chloride, 99%

Catalog No. p-199663
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232230050 5 g Glass bottle
AC232230500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232230050 Supplier Thermo Scientific Chemicals Supplier No. 232230050
Only null left
Add to Cart

CAS: 39098-97-0 | C6H5ClOS | 160.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39098-97-0
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005456
InChI Key AJYXPNIENRLELY-UHFFFAOYSA-N
Synonym 2-thiopheneacetyl chloride, 2-thiophen-2-yl acetyl chloride, 2-thienylacetyl chloride, 2-thiophene acetyl chloride, thiophene-2-acetyl chloride, 2-2-thienyl acetyl chloride, thiopheneacetyl chloride, 2-thiopheneacetylchloride, thienylacetyl chloride, thien-2-ylacetyl chloride
PubChem CID 162362
IUPAC Name 2-thiophen-2-ylacetyl chloride
SMILES C1=CSC(=C1)CC(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown
Density 1.3030g/mL
Boiling Point 130°C to 135°C (90.0 mmHg)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.55
Quantity 5 g
Specific Gravity 1.303
Formula Weight 160.62
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Thiopheneacetyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 254-290-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.