Learn More
Strem, An Ascensus Company CAS 6165-68-0. 25g. 2-Thiopheneboronic acid min. 97%. MFCD00151850
SDP
Catalog No. 502110685
Encompass
CAS 6165-68-0. 25g. 2-Thiopheneboronic acid min. 97%. MFCD00151850. Molecular Weight 127.96. Molecular Formula C4H3SB(OH)2. Color/form white to off-white pwdr. Strem 05-1040. http//www.strem.com/catalog/v/05-1040/
Catalog No. 50-211-0685 Supplier Strem, An Ascensus Company Supplier No. 05104025G
CAS 6165-68-0. 25g. 2-Thiopheneboronic acid min. 97%. MFCD00151850. Molecular Weight 127.96. Molecular Formula C4H3SB(OH)2. Color/form white to off-white pwdr. Strem 05-1040. http//www.strem.com/catalog/v/05-1040/