Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thiophenecarbonitrile

Catalog No. 15438510G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-438-510G 10 g
15-438-550G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-438-510G Supplier Sigma Aldrich Supplier No. 15438510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-thiophenecarbonitrile (2-cyanothiophene) was used in the preparation of thiaplatinacycles
  • It was also used in the synthesis of 2,2α-thienylpyrroles

Specifications

CAS 1003-31-2
Density 1.172 g/mL (at 25°C (literature))
Boiling Point 192°C (lit.)
Molecular Formula C5H3NS
Refractive Index n20/D 1.563 (literature)
Linear Formula C5H3NS
Quantity 10 g
MDL Number MFCD00005416
Synonym 2-Cyanothiophene; Thiophene-2-carbonitrile
Molecular Weight (g/mol) 109.15
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.