Learn More
Medchemexpress LLC 2-Thiophenemethanol | 636-72-6 | 5454 | 99.8% | 114.17 | 500 G
SDP
Catalog No. 5000404541
Encompass_Preferred
2-Thiophenemethanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.
  • Can be stored at 4°C, protected from light.
  • Stable in solvent at -80°C for 6 months.
  • Stable in solvent at -20°C for 1 month.
  • Forms clear solutions in various solubility protocols.
Catalog No. 50-004-04541 Supplier Medchemexpress LLC Supplier No. HY34751500G
2-Thiophenemethanol is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only.
  • Can be stored at 4°C, protected from light.
  • Stable in solvent at -80°C for 6 months.
  • Stable in solvent at -20°C for 1 month.
  • Forms clear solutions in various solubility protocols.