Learn More
Medchemexpress LLC 2-Thiophenemethanol | 636-72-6 | 98.6% | 114.17 | 25 G
SDP
Catalog No. 5000402513
Encompass_Preferred
2-Thiophenemethanol is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Can be used as a biological material or organic compound for life science related research
  • Intended for research use only
  • Appears as a liquid
  • Colorless to light yellow
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-02513 Supplier Medchemexpress LLC Supplier No. 25GHY34751
2-Thiophenemethanol is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only.
  • Can be used as a biological material or organic compound for life science related research
  • Intended for research use only
  • Appears as a liquid
  • Colorless to light yellow
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light