Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiophenesulfonyl chloride, 98%

Catalog No. AC420890250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420890250 25 g
AC420890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420890250 Supplier Thermo Scientific Chemicals Supplier No. 420890250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 16629-19-9 | C4H3ClO2S2 | 182.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16629-19-9
Molecular Formula C4H3ClO2S2
Molecular Weight (g/mol) 182.64
MDL Number MFCD00005426
InChI Key VNNLHYZDXIBHKZ-UHFFFAOYSA-N
Synonym 2-thiophenesulfonyl chloride, 2-thienylsulfonyl chloride, thiophene-2-sulphonyl chloride, 2-thiophenesulfonylchloride, 2-thienylsulfonylchloride, 2-thiophene sulfonyl chloride, chloro-2-thienylsulfone, pubchem7737, acmc-209dtw, thiophenesulfonyl chloride
PubChem CID 85518
IUPAC Name thiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 27°C to 29°C
Boiling Point 80°C (1.0 mmHg)
Flash Point >113°C
Assay Percent Range 97.5% min. (Ex Cl) (Argentometry)
Packaging Glass Bottle
Refractive Index 1.5722 to 1.5742
Quantity 25 g
Formula Weight 182.65
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Thiophenesulfonyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H StatementCauses severe skin burns and eye damage.Contact with water liberates toxic gas.
Precautionary Statement GHS P StatementIF SWALLOWED: rinse mouth.Do NOT induce vomiting.Wear eye protection/face protection.IF IN EYES: Rinse cautiously with water for several minutes.Remove contact lenses,if present and easy to do.Continue rinsi
EINECSNumber 240-677-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.