Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Thiophenesulfonyl fluoride

Catalog No. ALD001601G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD001601G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD001601G Supplier Sigma Aldrich Supplier No. ALD001601G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iodocyclopentane may be used in the synthesis of 4-chloro-3α-((2-cyclopentyl-1-oxo-1,2,3,4-tetrahydroisoquinolin-6- yloxy)methyl)biphenyl-3-carboxylic acid and N-(3-methoxyphenethyl)cyclopentanamine.

Specifications

CAS 382-99-0
Density 1.437 (at 25°C)
Molecular Formula C4H3FO2S2
Refractive Index n/D 1.509
Linear Formula C4H3FO2S2
MDL Number MFCD11109213
Quantity 25 mL
Synonym 2-Thiophenesulfonyl fluoride
Molecular Weight (g/mol) 166.19
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.