Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Thiouracil | 141-90-2 | 99.9% | 1 ML
SDP

Catalog No. 5000409484
Encompass_Preferred

2-Thiouracil (Thiouracil) is identified as a selective neuronal nitric oxide synthase (nNOS) inhibitor, with a Ki value of 20 μM. This compound is known to antagonize BH4-induced nNOS dimerization. Additionally, 2-Thiouracil functions as an antithyroid compound and a highly specific melanoma detector, and it has been observed to stimulate the growth of pea and corn root segments.

  • Selective neuronal nitric oxide synthase (nNOS) inhibitor (Ki value of 20 μM).
  • Antagonizes BH4-induced nNOS dimerization.
  • Functions as an antithyroid compound.
  • Acts as a highly specific melanoma detector.
  • Stimulates the growth of pea and corn root segments.
  • Purity: 99.9%.
  • Appearance: Solid, white to off-white.

Catalog No. 50-004-09484 Supplier Medchemexpress LLC Supplier No. HYB050310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Thiouracil (Thiouracil) is identified as a selective neuronal nitric oxide synthase (nNOS) inhibitor, with a Ki value of 20 μM. This compound is known to antagonize BH4-induced nNOS dimerization. Additionally, 2-Thiouracil functions as an antithyroid compound and a highly specific melanoma detector, and it has been observed to stimulate the growth of pea and corn root segments.

  • Selective neuronal nitric oxide synthase (nNOS) inhibitor (Ki value of 20 μM).
  • Antagonizes BH4-induced nNOS dimerization.
  • Functions as an antithyroid compound.
  • Acts as a highly specific melanoma detector.
  • Stimulates the growth of pea and corn root segments.
  • Purity: 99.9%.
  • Appearance: Solid, white to off-white.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.