Learn More
Medchemexpress LLC 2-Thiouracil | 141-90-2 | 99.9% | 5 G
SDP
Catalog No. 5000405275
Encompass_Preferred
2-Thiouracil (Thiouracil) is a selective neuronal nitric oxide synthase (nNOS) inhibitor with a Ki value of 20 μM. It antagonizes BH4-induced nNOS dimerization and has been observed to stimulate the growth of pea and corn root segments. Additionally, it functions as an antithyroid compound and a highly specific melanoma detector.
  • Functions as a selective neuronal nitric oxide synthase (nNOS) inhibitor
  • Antagonizes BH4-induced nNOS dimerization
  • Acts as an antithyroid compound
  • Serves as a highly specific melanoma detector
  • Stimulates growth of pea and corn root segments
Catalog No. 50-004-05275 Supplier Medchemexpress LLC Supplier No. HYB05035G
2-Thiouracil (Thiouracil) is a selective neuronal nitric oxide synthase (nNOS) inhibitor with a Ki value of 20 μM. It antagonizes BH4-induced nNOS dimerization and has been observed to stimulate the growth of pea and corn root segments. Additionally, it functions as an antithyroid compound and a highly specific melanoma detector.
  • Functions as a selective neuronal nitric oxide synthase (nNOS) inhibitor
  • Antagonizes BH4-induced nNOS dimerization
  • Acts as an antithyroid compound
  • Serves as a highly specific melanoma detector
  • Stimulates growth of pea and corn root segments