Learn More
Cayman Chemical 2-Thiouracil, Purity: ≥98%, CAS: 141-90-2, 250 g
SDP
Catalog No. 502910226
Encompass_Preferred
An antithyroid agent and a melanoma seeker; reduces sodium iodide-induced increases in thyroid weight in a mouse model of goiter at 2.5 mg/animal per day; preferentially accumulates into tumor cell melanin in a mouse model of melanoma; inhibits nNOS (Ki = 20 µM)
Catalog No. 50-291-0226 Supplier Cayman Chemical Supplier No. 34747250 G
An antithyroid agent and a melanoma seeker; reduces sodium iodide-induced increases in thyroid weight in a mouse model of goiter at 2.5 mg/animal per day; preferentially accumulates into tumor cell melanin in a mouse model of melanoma; inhibits nNOS (Ki = 20 µM)