Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Thiouracil 98.0+%, TCI America™
SDP

Catalog No. T022425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T022425G 25 g
T0224250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T022425G Supplier TCI America Supplier No. T022425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141-90-2
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00006039
InChI Key ZEMGGZBWXRYJHK-UHFFFAOYSA-N
Synonym 2-thiouracil, thiouracil, 4-hydroxy-2-mercaptopyrimidine, antagothyroid, antagothyroil, deracil, nobilen, 2-thioxo-2,3-dihydropyrimidin-4 1h-one, 2-mercaptopyrimidin-4-ol, 2-mercapto-4-hydroxypyrimidine
PubChem CID 1269845
ChEBI CHEBI:348530
IUPAC Name 2-sulfanylidene-1,2,3,4-tetrahydropyrimidin-4-one
SMILES O=C1NC(=S)NC=C1

Specifications

Melting Point 331°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 128.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiouracil
EINECSNumber (5)-0925
RTECSNumber YR1575000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.