Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Thiouracil (Thiouracil) | 141-90-2 | 99.9% | 25 G
SDP

Catalog No. 5000406577
Encompass_Preferred

2-Thiouracil (Thiouracil) is a selective neuronal nitric oxide synthase (nNOS) inhibitor with a Ki value of 20 μM. It antagonizes BH4-induced nNOS dimerization and acts as an antithyroid compound. This substance is also a highly specific melanoma detector and stimulates the growth of pea and corn root segments.

  • Selective neuronal nitric oxide synthase (nNOS) inhibitor
  • Antagonizes BH4-induced nNOS dimerization
  • Acts as an antithyroid compound
  • Highly specific melanoma detector
  • Stimulates the growth of pea and corn root segments
  • Molecular weight: 128.16
  • Formula: C4H4N2OS
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-004-06577 Supplier Medchemexpress LLC Supplier No. 25GHYB0503
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Thiouracil (Thiouracil) is a selective neuronal nitric oxide synthase (nNOS) inhibitor with a Ki value of 20 μM. It antagonizes BH4-induced nNOS dimerization and acts as an antithyroid compound. This substance is also a highly specific melanoma detector and stimulates the growth of pea and corn root segments.

  • Selective neuronal nitric oxide synthase (nNOS) inhibitor
  • Antagonizes BH4-induced nNOS dimerization
  • Acts as an antithyroid compound
  • Highly specific melanoma detector
  • Stimulates the growth of pea and corn root segments
  • Molecular weight: 128.16
  • Formula: C4H4N2OS
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.