Learn More
Cayman Chemical 2-Thiouridine, Purity: ≥98%, CAS: 20235-78-3, 50 mg
SDP
Catalog No. 502983602
Encompass_Preferred
A modified nucleobase found in tRNAs that is known to stabilize U:A pairs
Catalog No. 50-298-3602 Supplier Cayman Chemical Supplier No. 2101150 MG
A modified nucleobase found in tRNAs that is known to stabilize U:A pairs