Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-thiouridine | 20235-78-3 | MFCD03788697 | 99.7% | 260.27 g/mol | C9H12N2O5S | 1 ML
SDP

Catalog No. 5000226548
Encompass_Preferred

2-Thiouridine is a modified nucleoside provided as a ready-to-use 10 mM solution in DMSO for biochemical and virology research, used to study RNA base-pairing, nonenzymatic replication, and antiviral activity.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied volume 1 mL.
  • Reported purity 99.7%.
  • Molecular weight 260.27 g/mol.
  • CAS number 20235-78-3.
  • In-solution storage: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-26548 Supplier Medchemexpress LLC Supplier No. HY11781310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Thiouridine is a modified nucleoside provided as a ready-to-use 10 mM solution in DMSO for biochemical and virology research, used to study RNA base-pairing, nonenzymatic replication, and antiviral activity.

  • Ready-to-use 10 mM solution in DMSO.
  • Supplied volume 1 mL.
  • Reported purity 99.7%.
  • Molecular weight 260.27 g/mol.
  • CAS number 20235-78-3.
  • In-solution storage: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.