Learn More
Medchemexpress LLC 2-Tolylboronic acid | 16419-60-6 | 135.96 | 25 G
SDP
Catalog No. 5000449963
Encompass_Preferred
2-Tolylboronic acid is a biochemical analysis reagent and an important intermediate. It can be used for the synthesis of other active compounds and is suitable for research use only.
  • Biochemical analysis reagent
  • Important intermediate for organic synthesis
  • Substrate in Suzuki cross-coupling reactions
  • Can be used to evaluate catalyst activity
Catalog No. 50-004-49963 Supplier Medchemexpress LLC Supplier No. 25GHYW001232
2-Tolylboronic acid is a biochemical analysis reagent and an important intermediate. It can be used for the synthesis of other active compounds and is suitable for research use only.
  • Biochemical analysis reagent
  • Important intermediate for organic synthesis
  • Substrate in Suzuki cross-coupling reactions
  • Can be used to evaluate catalyst activity