Learn More
Perkin Elmer US LLC 2 Ton Hydraulic KBr Press
Catalog No. 502113261
Encompass_Preferred
With its innovative sealed hydraulic unit and durable design, this benchtop press is made for daily use and maintenance-free operation. It requires, but does not include, the 7 mm Pellet Die and Ring Holder Assembly (L1272492) and the KBr Disc Holder with Rectangular Mount (L1272493)
Catalog No. 50-211-3261 Supplier Perkin Elmer US LLC Supplier No. L1272491
With its innovative sealed hydraulic unit and durable design, this benchtop press is made for daily use and maintenance-free operation. It requires, but does not include, the 7 mm Pellet Die and Ring Holder Assembly (L1272492) and the KBr Disc Holder with Rectangular Mount (L1272493)