Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[trans-4-(Boc-amino)cyclohexyl]ethylamine, 97%, Thermo Scientific™

Catalog No. AAH62326MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62326MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62326MD Supplier Thermo Scientific Chemicals Supplier No. H62326MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1212272-05-3
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03844600
InChI Key SSZKBGFWNFAXNC-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
PubChem CID 24720893
IUPAC Name tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCN

Specifications

Quantity 250 mg
Formula Weight 242.35
Percent Purity 97%
Chemical Name or Material 2-[trans-4-(Boc-amino)cyclohexyl]ethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.