Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Tribromomethylquinoline 98.0+%, TCI America™
SDP

Catalog No. T14535G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T14535G Supplier TCI America Supplier No. T14535G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 613-53-6
Molecular Formula C10H6Br3N
Molecular Weight (g/mol) 379.88
MDL Number MFCD00006755
InChI Key UDYYQHILRSDDMP-UHFFFAOYSA-N
Synonym 2-tribromomethyl quinoline, quinoline, 2-tribromomethyl, alpha,alpha,alpha-tribromoquinaldine, 2-alpha,alpha,alpha-tribromomethyl quinoline, .alpha.,.alpha.,.alpha.-tribromoquinaldine, acmc-209mrr, 2-tribrommethyl-chinolin, 2-tribromo-methyl-quinoline, quinoline,2-tribromomethyl, udyyqhilrsddmp-uhfffaoysa
PubChem CID 69178
IUPAC Name 2-(tribromomethyl)quinoline
SMILES BrC(Br)(Br)C1=CC=C2C=CC=CC2=N1

Specifications

Melting Point 129°C
Quantity 5 g
Formula Weight 379.88
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Tribromomethylquinoline
RTECSNumber VC3250750
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.