Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Tributylstannyl)Pyrimidine

Catalog No. 7211741G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-117-41G 1 g
72-117-45G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 72-117-41G Supplier Sigma Aldrich Supplier No. 7211741G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(Tributylstannyl)pyrimidine is an organotin compound used in Stille coupling reaction. This can be used as one of the precursors in the synthesis of 2-aminopyridine oxazolidinones as potent and selective tankyrase (TNKS) inhibitors and the synthesis of canagliflozin, a novel inhibitor for sodium-dependent glucose cotransporter.It can also be used in the preparation of various (2-pyrimidyl)silanes.

Specifications

CAS 153435-63-3
Density 1.164 g/mL at 25°C
Molecular Formula C16H30N2Sn
Refractive Index n20/D 1.512
Linear Formula C16H30N2Sn
MDL Number MFCD01319085
Quantity 1 g
Synonym 2-Tributylstannanylpyrimidine; Tributyl (2-pyrimidyl)stannane
Molecular Weight (g/mol) 369.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.