Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Tributylstannylthiazole

Catalog No. 6425415G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-254-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 64-254-15G Supplier Sigma Aldrich Supplier No. 6425415G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent for arylation of thiazole by Stille cross-coupling.

Specifications

CAS 101469-92-5
Melting Point 60°C to 64°C
Molecular Formula C9H17NO3
Linear Formula C9H17NO3
Refractive Index n20/D 1.5200
MDL Number MFCD01317839
Quantity 5 g
Synonym (S)-N-(tert-Butoxycarbonyl)-(+)-3-pyrrolidinol
Molecular Weight (g/mol) 187.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.