Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Tridecanone | 593-08-8 | MFCD00008968 | 99.94% | 198.35 | 500 MG
SDP

Catalog No. 5000453727
Encompass_Preferred

2-Tridecanone is a nonalkaloid insecticide isolated from the wild tomato Lycopersicon hirsutum f. glabratum. It is also a volatile organic compound. This compound is classified under ketones, aldehydes, and acids, and its initial source is plants from other families.

  • Molecular weight is 198.35
  • Chemical formula is C13H26O
  • Appears as a solid below 24°C and a liquid above 27°C
  • Color is white to off-white
  • Classified as a ketone, aldehyde, or acid
  • Derived from plants
  • For research use only

Catalog No. 50-004-53727 Supplier Medchemexpress LLC Supplier No. HYW009811500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Tridecanone is a nonalkaloid insecticide isolated from the wild tomato Lycopersicon hirsutum f. glabratum. It is also a volatile organic compound. This compound is classified under ketones, aldehydes, and acids, and its initial source is plants from other families.

  • Molecular weight is 198.35
  • Chemical formula is C13H26O
  • Appears as a solid below 24°C and a liquid above 27°C
  • Color is white to off-white
  • Classified as a ketone, aldehyde, or acid
  • Derived from plants
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.