Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-[(Triethylsilyl)ethynyl]pyridine

Catalog No. RNI001481G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RNI001481G 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RNI001481G Supplier Sigma Aldrich Supplier No. RNI001481G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The infrared spectra of solid, gaseous and liquid and solid trans 3-pentenenitrile were studied.trans-3-Pentenenitrile was used to study the sensory and motor conduction velocities (SCV and MCV) and amplitudes of the motor and sensory action potentials (AMAP and ASAP) of the tail nerve in male Sprague-Dawley rats.DuPont product

Specifications

Molecular Formula C13H19NSi
Linear Formula C13H19NSi
MDL Number MFCD27978291
Quantity 250 mL
Molecular Weight (g/mol) 217.38
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.