Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)aniline, 97%, Thermo Scientific™

Catalog No. SB01054EB
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01054EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01054EB Supplier Thermo Scientific Chemicals Supplier No. SB01054EB

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00035959
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CF3OC6H4NH2
Quantity 25 g
Formula Weight 177.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethoxy)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.