Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. T23715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23715G Supplier TCI America Supplier No. T23715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63968-85-4
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.121
MDL Number MFCD00042409
InChI Key ACNBBQGAWMHXLA-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzonitrile, o-trifluoromethoxybenzonitrile, benzonitrile, 2-trifluoromethoxy, 2-trifluoromethoxy benzenecarbonitrile, pubchem4824, acmc-1b9l5, ksc493e9p, o-trifluoromethoxy benzonitrile, acnbbqgawmhxla-uhfffaoysa
PubChem CID 2777232
IUPAC Name 2-(trifluoromethoxy)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)OC(F)(F)F

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
UN Number 3276
Formula Weight 187.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.