Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)benzyl Alcohol 96.0+%, TCI America™
SDP

Catalog No. T25935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25935G 5 g
T25931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25935G Supplier TCI America Supplier No. T25935G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175278-07-6
Molecular Formula C8H7F3O2
Molecular Weight (g/mol) 192.14
MDL Number MFCD00153285
InChI Key ICOVMLDFMWLRJO-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzyl alcohol, 2-trifluoromethoxy phenyl methanol, 2-trifluoromethoxy benzylalcohol, 2-trifluoromethoxybenzyl alcohol, benzenemethanol, 2-trifluoromethoxy, 2-trifluoromethoxy phenyl methan-1-ol, pubchem2745, acmc-209e9m, rarechem al bd 0282
PubChem CID 2777245
IUPAC Name [2-(trifluoromethoxy)phenyl]methanol
SMILES OCC1=CC=CC=C1OC(F)(F)F

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
Formula Weight 192.14
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)benzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.