Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)phenylacetonitrile 95.0+%, TCI America™
SDP

Catalog No. T23655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T23655G Supplier TCI America Supplier No. T23655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 137218-25-8
Molecular Formula C9H6F3NO
Molecular Weight (g/mol) 201.15
MDL Number MFCD00236326
InChI Key OIKWCWVMUBCXJM-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy phenylacetonitrile, 2-triflnoromethoxybenzyl cyanide, 2-2-trifluoromethoxy phenyl acetonitrile, 2-trifluoromethoxybenzyl cyanide, 1 2-trifluoromethoxy phenyl acetonitrile, 2-trifluoromethoxy benzyl cyanide, 2-trifluoromethoxybenzylcyanide, 2-trifluoromethoxy-phenyl-acetonitrile, benzeneacetonitrile, 2-trifluoromethoxy
PubChem CID 2777321
IUPAC Name 2-[2-(trifluoromethoxy)phenyl]acetonitrile
SMILES FC(F)(F)OC1=CC=CC=C1CC#N

Specifications

Color Yellow
Boiling Point 223°C
Quantity 5 g
UN Number 3276
Formula Weight 201.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)phenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.