Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Trifluoromethoxy)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. T24135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2413-5G 5 g
T24131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2413-5G Supplier TCI America Supplier No. T24135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175676-65-0
Molecular Formula C7H6BF3O3
Molecular Weight (g/mol) 205.93
MDL Number MFCD01320763
InChI Key AIJCNTOYZPKURP-UHFFFAOYSA-N
PubChem CID 2777204
IUPAC Name [2-(trifluoromethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1OC(F)(F)F

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 5 g
Synonym 2-trifluoromethoxy phenylboronic acid, 2-trifluoromethoxyphenylboronic acid, 2-trifluormethoxy phenylboronic acid, 2-trifluoromethoxy benzeneboronic acid, 2-trifluoromethoxy phenyl boronic acid
Formula Weight 205.93
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.