Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(Trifluoromethyl)benzaldehyde

Catalog No. 250694100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-069-4100G 100 g
25-069-425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-069-4100G Supplier Sigma Aldrich Supplier No. 250694100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-(trifluoromethyl)benzaldehyde has been used in the preparation of:
  • -d-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates
  • Methyl (e)-2-[2-[(e)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via wittig-horner reaction

Specifications

CAS 447-61-0
Density 1.32 g/mL (at 25°C (literature))
Molecular Formula C8H5F3O
Linear Formula CF3C6H4CHO
Refractive Index n20/D 1.466 (literature)
MDL Number MFCD00003337
Quantity 100 g
Synonym alpha,alpha,alpha-Trifluoro-o-tolualdehyde
Molecular Weight (g/mol) 174.12
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.